siRNA Details
| Gene Symbol: D830032E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102307.1 | 2 | 172 | |||
| GenBank mRNA | AK081398.1 | - | 80 | |||
| AK085943.1 | - | 2 | 147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D830032E09Rik| Gene Symbol: D830032E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102307.1 | 2 | 172 | |||
| GenBank mRNA | AK081398.1 | - | 80 | |||
| AK085943.1 | - | 2 | 147 | |||