siRNA Details
| Gene Symbol: Gm26515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131823.1 | - | 2 | 459 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707603.1 | - | 2 | 454 | Chr. 5 - 110779886 - 110780861 [+] on Build GRCm38 | |

Gm26515
Genomic Map