siRNA Details
| Gene Symbol: 5830454J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018012.1 | - | 1 | 133 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717943.1 | - | 1 | 131 | Chr. 17 - 28695103 - 28695753 [+] on Build GRCm38 | |

5830454J16Rik
Genomic Map