siRNA Details
| Gene Symbol: Chmp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008733.1 | - | 486 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chmp4c| Gene Symbol: Chmp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008733.1 | - | 486 | |||