siRNA Details
| Gene Symbol: 4933437F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017098.1 | - | 1 | 1374 | ||
| AV283813.1 | - | 1 | 142 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933437F24Rik| Gene Symbol: 4933437F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017098.1 | - | 1 | 1374 | ||
| AV283813.1 | - | 1 | 142 | |||