siRNA Details
| Gene Symbol: Gm20757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046027.1 | 8 | 1067 | |||
| GenBank mRNA | AK053604.1 | - | 8 | 1068 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB541330.2 | - | 2 | 153 | Chr. 10 - 92171301 - 92257192 [-] on Build GRCm38 | |

Gm20757
Genomic Map