siRNA Details
| Gene Symbol: 4930556C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040506.1 | 4 | 880 | |||
| GenBank mRNA | AK016142.1 | - | 4 | 881 | ||
| BB020005.1 | - | 1 | 140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930556C24Rik| Gene Symbol: 4930556C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040506.1 | 4 | 880 | |||
| GenBank mRNA | AK016142.1 | - | 4 | 881 | ||
| BB020005.1 | - | 1 | 140 | |||