siRNA Details
| Gene Symbol: 4930556J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045714.1 | 2 | 171 | |||
| GenBank mRNA | AK016147.1 | - | 2 | 172 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930556J02Rik| Gene Symbol: 4930556J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045714.1 | 2 | 171 | |||
| GenBank mRNA | AK016147.1 | - | 2 | 172 | ||