siRNA Details
| Gene Symbol: 4930556J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045714.1 | 3 | 576 | |||
| GenBank mRNA | AK016147.1 | - | 3 | 577 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU945712.1 | - | 2 | 247 | Chr. 14 - 61493334 - 61500377 [+] on Build GRCm38 | |

4930556J02Rik
Genomic Map