siRNA Details
| Gene Symbol: 5230401M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017231.1 | - | 1 | 1145 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5230401M06Rik| Gene Symbol: 5230401M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017231.1 | - | 1 | 1145 | ||