siRNA Details
| Gene Symbol: 9430051O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020465.1 | - | 1 | 379 | ||
| AK035689.1 | - | 1 | 865 | |||
| BB122864.2 | - | 1 | 177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9430051O21Rik| Gene Symbol: 9430051O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020465.1 | - | 1 | 379 | ||
| AK035689.1 | - | 1 | 865 | |||
| BB122864.2 | - | 1 | 177 | |||