siRNA Details
| Gene Symbol: C430049B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038184.1 | 4 | 2981 | |||
| NR_038185.1 | 4 | 2695 | ||||
| GenBank mRNA | AK020467.1 | - | 1 | 154 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C430049B03Rik| Gene Symbol: C430049B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038184.1 | 4 | 2981 | |||
| NR_038185.1 | 4 | 2695 | ||||
| GenBank mRNA | AK020467.1 | - | 1 | 154 | ||