siRNA Details
| Gene Symbol: 9430040K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020461.1 | - | 1 | 148 | ||
| AK142550.1 | - | 1 | 2575 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9430040K09Rik| Gene Symbol: 9430040K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020461.1 | - | 1 | 148 | ||
| AK142550.1 | - | 1 | 2575 | |||