siRNA Details
| Gene Symbol: Limk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042206.1 | - | 1 | 1225 | ||
| AK044582.1 | - | 1 | 1228 | |||
| AK161560.1 | - | 1 | 1215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Limk2| Gene Symbol: Limk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042206.1 | - | 1 | 1225 | ||
| AK044582.1 | - | 1 | 1228 | |||
| AK161560.1 | - | 1 | 1215 | |||