siRNA Details
| Gene Symbol: Limk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042206.1 | - | 1 | 678 | ||
| AK044582.1 | - | 1 | 681 | |||
| AK161560.1 | - | 1 | 668 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV327553.2 | - | 1 | 91 | Chr. 11 - 3407893 - 3408562 [-] on Build GRCm38 | |

Limk2
Genomic Map