siRNA Details
| Gene Symbol: 1700022H01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006239.1 | - | 2 | 182 | ||
| AV046979.2 | - | 2 | 107 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700022H01Rik| Gene Symbol: 1700022H01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006239.1 | - | 2 | 182 | ||
| AV046979.2 | - | 2 | 107 | |||