siRNA Details
| Gene Symbol: 1700048C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006722.1 | - | 1 | 257 | ||
| AV045704.2 | - | 1 | 261 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706710.1 | - | 1 | 257 | Chr. 7 - 130398329 - 130398630 [+] on Build GRCm38 | |

1700048C15Rik
Genomic Map