siRNA Details
| Gene Symbol: 1700048B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006721.1 | - | 2 | 453 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB274787.1 | - | 2 | 220 | Chr. 13 - 50914040 - 50933349 [-] on Build GRCm38 | |

1700048B10Rik
Genomic Map