siRNA Details
| Gene Symbol: 1700047O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006720.1 | - | 1 | 204 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706709.1 | - | 1 | 214 | Chr. 7 - 12780850 - 12781602 [+] on Build GRCm38 | |

1700047O18Rik
Genomic Map