siRNA Details
| Gene Symbol: E430022K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088651.1 | - | 1 | 1645 | ||
| AK169489.1 | - | 1 | 1602 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E430022K19Rik| Gene Symbol: E430022K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088651.1 | - | 1 | 1645 | ||
| AK169489.1 | - | 1 | 1602 | |||