siRNA Details
| Gene Symbol: 9130223C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020287.1 | - | 1 | 299 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9130223C08Rik| Gene Symbol: 9130223C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020287.1 | - | 1 | 299 | ||