siRNA Details
| Gene Symbol: 9130223C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020287.1 | - | 1 | 108 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720865.1 | - | 1 | 108 | Chr. 4 - 53572440 - 53573037 [+] on Build GRCm38 | |

9130223C08Rik
Genomic Map