siRNA Details
| Gene Symbol: Dpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019051.1 | - | 2 | 353 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708591.1 | - | 2 | 351 | Chr. 19 - 5911826 - 5912809 [-] on Build GRCm38 | |

Dpf2
Genomic Map