siRNA Details
| Gene Symbol: Gm15247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019053.1 | - | 2 | 317 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708654.1 | - | 2 | 316 | Chr. X - 169985801 - 169986940 [-] on Build GRCm38 | |

Gm15247
Genomic Map