siRNA Details
| Gene Symbol: Gm16226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK160425.1 | - | 164 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707211.1 | - | 2 | 162 | Chr. 5 - 99682441 - 99715770 [+] on Build GRCm38 | |

Gm16226
Genomic Map