siRNA Details
| Gene Symbol: Zfp141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137863.1 | - | 1 | 629 | ||
| BC063263.1 | - | 7 | 3835 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB155738.1 | - | 1 | 33 | Chr. 7 - 42473389 - 42473675 [-] on Build GRCm38 | |

Zfp141
Genomic Map