siRNA Details
Gene Symbol: Zfp141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137863.1 | - | 1 | 765 | ||
BC063263.1 | - | 7 | 3971 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB155738.1 | - | 1 | 169 | Chr. 7 - 42473389 - 42473675 [-] on Build GRCm38 |
