siRNA Details
| Gene Symbol: Ago3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031725.1 | - | 1 | 566 | ||
| AK047638.1 | - | 1 | 3736 | |||
| AK081408.1 | - | 1 | 527 | |||
| BC060127.1 | - | 1 | 3273 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ago3| Gene Symbol: Ago3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031725.1 | - | 1 | 566 | ||
| AK047638.1 | - | 1 | 3736 | |||
| AK081408.1 | - | 1 | 527 | |||
| BC060127.1 | - | 1 | 3273 | |||