siRNA Details
| Gene Symbol: 4930515B02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040654.1 | 1 | 144 | |||
| GenBank mRNA | AK015793.1 | - | 1 | 144 | ||
| BY715338.1 | - | 1 | 142 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930515B02Rik| Gene Symbol: 4930515B02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040654.1 | 1 | 144 | |||
| GenBank mRNA | AK015793.1 | - | 1 | 144 | ||
| BY715338.1 | - | 1 | 142 | |||