siRNA Details
Gene Symbol: 4930515B02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040654.1 | 2 | 233 | |||
GenBank mRNA | AK015793.1 | - | 2 | 233 | ||
BY715338.1 | - | 2 | 231 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930515B02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040654.1 | 2 | 233 | |||
GenBank mRNA | AK015793.1 | - | 2 | 233 | ||
BY715338.1 | - | 2 | 231 |