siRNA Details
| Gene Symbol: 4930458D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015484.1 | - | 1 | 760 | ||
| AK036594.1 | - | 1 | 1816 | |||
| AK133807.1 | - | 1 | 752 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB028846.2 | - | 1 | 575 | Chr. 5 - 100841632 - 100845859 [-] on Build GRCm38 | |

4930458D05Rik
Genomic Map