siRNA Details
| Gene Symbol: 4930458D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015484.1 | - | 1 | 578 | ||
| AK036594.1 | - | 1 | 1634 | |||
| AK133807.1 | - | 1 | 570 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930458D05Rik| Gene Symbol: 4930458D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015484.1 | - | 1 | 578 | ||
| AK036594.1 | - | 1 | 1634 | |||
| AK133807.1 | - | 1 | 570 | |||