siRNA Details
Gene Symbol: 4930515L03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040632.1 | 2, 3 | 157 | |||
GenBank mRNA | AK015798.1 | - | 2, 3 | 158 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930515L03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040632.1 | 2, 3 | 157 | |||
GenBank mRNA | AK015798.1 | - | 2, 3 | 158 |