siRNA Details
| Gene Symbol: 4930515L03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040632.1 | 3 | 196 | |||
| GenBank mRNA | AK015798.1 | - | 3 | 197 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930515L03Rik| Gene Symbol: 4930515L03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040632.1 | 3 | 196 | |||
| GenBank mRNA | AK015798.1 | - | 3 | 197 | ||