siRNA Details
Gene Symbol: 4930515L03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040632.1 | 4 | 426 | |||
GenBank mRNA | AK015798.1 | - | 4 | 427 | ||
AV266265.1 | - | 1 | 126 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930515L03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040632.1 | 4 | 426 | |||
GenBank mRNA | AK015798.1 | - | 4 | 427 | ||
AV266265.1 | - | 1 | 126 |