siRNA Details
| Gene Symbol: 4921508D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045802.1 | 4 | 725 | |||
| GenBank mRNA | AK014842.1 | - | 4 | 727 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921508D12Rik| Gene Symbol: 4921508D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045802.1 | 4 | 725 | |||
| GenBank mRNA | AK014842.1 | - | 4 | 727 | ||