siRNA Details
| Gene Symbol: Gm38414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082764.1 | - | 2 | 239 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB651724.1 | - | 2 | 237 | Chr. 8 - 36156012 - 36160194 [+] on Build GRCm38 | |

Gm38414
Genomic Map