siRNA Details
| Gene Symbol: Gm38407 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110487.1 | 1 | 72 | |||
| GenBank mRNA | AK029853.1 | - | 1 | 74 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB616152.1 | - | 1 | 74 | Chr. 12 - 10446388 - 10467795 [+] on Build GRCm38 | |

Gm38407
Genomic Map