siRNA Details
| Gene Symbol: Gm38407 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110487.1 | 9 | 1738 | |||
| GenBank mRNA | AK029853.1 | - | 9 | 1740 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm38407| Gene Symbol: Gm38407 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110487.1 | 9 | 1738 | |||
| GenBank mRNA | AK029853.1 | - | 9 | 1740 | ||