siRNA Details
| Gene Symbol: Gm2762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037991.1 | 8 | 1091 | |||
| GenBank mRNA | AK006343.1 | - | 8 | 1069 | ||
| AK030238.1 | - | 8 | 1092 | |||
| BC100495.1 | - | 8 | 1072 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm2762| Gene Symbol: Gm2762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037991.1 | 8 | 1091 | |||
| GenBank mRNA | AK006343.1 | - | 8 | 1069 | ||
| AK030238.1 | - | 8 | 1092 | |||
| BC100495.1 | - | 8 | 1072 | |||