siRNA Details
| Gene Symbol: 3110040K02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014159.1 | - | 1 | 423 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY713134.1 | - | 1 | 423 | Chr. 2 - 158411515 - 158412002 [+] on Build GRCm38 | |

3110040K02Rik
Genomic Map