siRNA Details
| Gene Symbol: 1700055K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006845.1 | - | 1 | 536 | ||
| AK018863.1 | - | 1 | 535 | |||
| AK018893.1 | - | 1 | 536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700055K11Rik| Gene Symbol: 1700055K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006845.1 | - | 1 | 536 | ||
| AK018863.1 | - | 1 | 535 | |||
| AK018893.1 | - | 1 | 536 | |||