siRNA Details
| Gene Symbol: 2310057M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014156.1 | - | 1 | 538 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY713132.1 | - | 1 | 538 | Chr. 7 - 131355166 - 131355742 [-] on Build GRCm38 | |

2310057M21Rik
Genomic Map