siRNA Details
| Gene Symbol: 2310057M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014156.1 | - | 1 | 1130 | ||
| AK076241.1 | - | 1 | 26 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310057M21Rik| Gene Symbol: 2310057M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014156.1 | - | 1 | 1130 | ||
| AK076241.1 | - | 1 | 26 | |||