siRNA Details
| Gene Symbol: 2610202C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011859.1 | - | 1 | 443 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711078.1 | - | 1 | 440 | Chr. 10 - 58284431 - 58285087 [+] on Build GRCm38 | |

2610202C22Rik
Genomic Map