siRNA Details
| Gene Symbol: 2610201A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA222883.1 | - | 1 | 12 | ||
| AA473299.1 | - | 1 | 12 | |||
| AK011855.1 | - | 1 | 597 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610201A13Rik| Gene Symbol: 2610201A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA222883.1 | - | 1 | 12 | ||
| AA473299.1 | - | 1 | 12 | |||
| AK011855.1 | - | 1 | 597 | |||