siRNA Details
| Gene Symbol: 2610201A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011855.1 | - | 1 | 293 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM936630.1 | - | 1 | 56 | Chr. 11 - 33150573 - 33151207 [-] on Build GRCm38 | |

2610201A13Rik
Genomic Map