siRNA Details
| Gene Symbol: Gm16863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046162.1 | 5 | 2252 | |||
| GenBank mRNA | AK043964.1 | - | 5 | 2253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16863| Gene Symbol: Gm16863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046162.1 | 5 | 2252 | |||
| GenBank mRNA | AK043964.1 | - | 5 | 2253 | ||