siRNA Details
| Gene Symbol: A730006G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110485.1 | 3 | 1607 | |||
| GenBank mRNA | AK042565.1 | - | 3 | 1608 | ||
| AK043085.1 | - | 3 | 1608 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A730006G06Rik| Gene Symbol: A730006G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110485.1 | 3 | 1607 | |||
| GenBank mRNA | AK042565.1 | - | 3 | 1608 | ||
| AK043085.1 | - | 3 | 1608 | |||