siRNA Details
| Gene Symbol: A730006G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110485.1 | 3 | 573 | |||
| GenBank mRNA | AK042565.1 | - | 3 | 574 | ||
| AK043085.1 | - | 3 | 574 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB639674.1 | - | 3 | 571 | Chr. 17 - 48065967 - 48090376 [-] on Build GRCm38 | |

A730006G06Rik
Genomic Map